missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807035
Additional Details : Weight : 0.05000kg
Description
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.Specifications
CITED1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q99966 | |
CITED1 | |
Full length human recombit protein of human CITED1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI1C6 | |
Unconjugated | |
CITED1 | |
MSG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4435 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |