missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Linear Formula | HOC(COOH)(CH2COOH)2 |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol |
Formula Weight | 192.12 |
Percent Purity | 99.5 to 100.5% (based on anhydrous substance) |
Quantity | 6 x 1 kg |
Explosion Limit | 8% (65°F) |
Residue after Ignition | ≤0.05% (as SO4) |
Chemical Name or Material | Citric Acid, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling

Product Identifier
- Citric acid
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Citric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction