Learn More
Scharlab CITRONSYRA-1-HYDRAT PA 500G
Brand:  Scharlab AC07250500
CITRONSYRA-1-HYDRAT PA 500G
Product Code. 11369933
  • 44.45 EUR / 500g
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Citric acid monohydrate, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Scharlab CITRONSYRA-1-HYDRAT PA 500G >