Learn More
Emma Safety Footwear Clay S3 Shoe
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.
Brand: Emma Safety Footwear MM98054942
This item is not returnable.
View return policy
Description
- S3-Safetyshoe with sole plate for wet work
- Metal Free
- The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
- It gets its name from the highly supportive reinforced heel.
- This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
- DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
- Industries:
- Logistics & transport
- Electronics
- Construction & technology
- Manufacturing
- Nature & environment
Specifications
ISO 20345:2011 | |
Black | |
Hydro-Tec Antibacterial | |
42 | |
Low-cut |
Laces | |
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment | |
Nubuck, Oiled Nubuck | |
Standard Width | |
Safety Shoe |
Product Suggestions
Customers who viewed this item also viewed
CE_17512636.pdf
File size: 325.6 kB
Product Content Correction
Product Title
Emma Safety Footwear Clay S3 Shoe > Safety Shoe; Size: 42
Spot an opportunity for improvement?Share a Content Correction