Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Brand:  Cole-Parmer 50108-81
 View more versions of this product
Product Code. 30052960
  • 11.70 EUR / Pack of 10
Estimated Shipment: 30-07-2025
Explore more special offers
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
10/Pk.
Hose Barb Reducer
Straight
-23°C
1/8 to 1/16 in.
121°C
Natural
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene > 1/8 to 1/16 in.