Learn More
BINDER™ CO2 Gassing Module
CO2 gassing module
3150.00 EUR - 3890.00 EUR
Specifications
Material | Stainless Steel |
---|---|
Type | Module |
Includes | NDIR CO2 Sensor and Analog Output 4 to 20 mA |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238252
|
BINDER™
8012-2138 |
KBF 470, KBF 720, KBF 1060, KBF 1600, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
3150.00 EUR
Each |
In Stock
Log in to see stock. |
||||||
30238333
|
BINDER™
8012-2140 |
KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600, KBF 470, KBF 720, KBF 1060, KBF 1600 |
3890.00 EUR
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
Description
- CO2 gassing module with NDIR CO2 sensor and analog output 4-20 mA
- Choose between 0 to 20% and 0 to 1%
Specifications
Stainless Steel | |
Module | |
NDIR CO2 Sensor and Analog Output 4 to 20 mA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ CO2 Gassing Module