Learn More
BINDER™ Coated Heat Exchanger
Heat exchanger that is coated in plastic to prevent damage.
Brand: BINDER™ 8012-2583
This item is not returnable.
View return policy
Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
KBF PRO 260 | |
Aluminum | |
Heat Exchanger |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BINDER™ Coated Heat Exchanger > For use with KBF PRO 260
Spot an opportunity for improvement?Share a Content Correction