Learn More
CombiCheck™ 18700 90 Multi-Parameter Standard Solution, WTW™
WTW™ CombiCheck™ Solutions are ready-to-use multi-parameter standards. Every package contains a standard solution and an addition solution. Both solutions can be used directly without dilution, for quality assurance purposes.
Brand:  WTW 252501
147.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16439475
  • 154.00 EUR / Each
Description
  • Ready-to-use multi-parameter photometric standards
  • The standard solution ensures the accuracy of the results from the entire system
  • The addition solution allows you to check sample-dependent influences
  • Up to 280 determinations are possible using the addition solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.250 mg/L Cd; 2.00 mg/L Cu; 1.00 mg/L Fe; 1.00 mg/L Mn
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
Product Title
CombiCheck™ 18700 90 Multi-Parameter Standard Solution, WTW™ > 0.250 mg/L Cd; 2.00 mg/L Cu; 1.00 mg/L Fe; 1.00 mg/L Mn