Learn More
CombiCheck™ 18700 90 Multi-Parameter Standard Solution, WTW™
WTW™ CombiCheck™ Solutions are ready-to-use multi-parameter standards. Every package contains a standard solution and an addition solution. Both solutions can be used directly without dilution, for quality assurance purposes.
Brand: WTW 252501
147.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ready-to-use multi-parameter photometric standards
- The standard solution ensures the accuracy of the results from the entire system
- The addition solution allows you to check sample-dependent influences
- Up to 280 determinations are possible using the addition solution
Specifications
0.250 mg/L Cd; 2.00 mg/L Cu; 1.00 mg/L Fe; 1.00 mg/L Mn |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CombiCheck 90
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, copper(II) nitrate
Product Content Correction
Product Title
CombiCheck™ 18700 90 Multi-Parameter Standard Solution, WTW™ > 0.250 mg/L Cd; 2.00 mg/L Cu; 1.00 mg/L Fe; 1.00 mg/L Mn
Spot an opportunity for improvement?Share a Content Correction