Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ MS15-20-2
 View more versions of this product
Product Code. 11843393
  • 39.55 EUR / Each
Estimated Shipment: 14-05-2025
Description
  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
20
2 mm
Comb
1 Each
43 ÎĽL
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Fisherbrand and Fisher BioReagents Focus on electrophoresis
Product Title
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick > Sample size: 43ÎĽL; Number of Wells: 20