Learn More
Kern Sohn Compact Laboratory Balance PCB
The standard balance ideal for use in the laboratory.
Brand: Kern Sohn PCB 1000-1
This item is not returnable.
View return policy
Description
- PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
- Freely programmable weighing unit, e.g. display direct in special units such as length of thread g/m, paper weight g/m2, or similar
- With the recipe function you can weigh the different ingredients of a mixture. As a check, you can also call up the total weight of all the ingredients
- Plus/minus weighing procedures
- Protective working cover included with delivery
Specifications
1000 g | |
0.2 g | |
Compact Precision Blanace | |
130 mm |
0.1 g | |
0.1 g | |
130 mm | |
Universal Mains Adapter |