Learn More
Kern Sohn Compact Laboratory Balance PCB
The standard balance ideal for use in the laboratory.
POA - 387.60 EUR
Specifications
Type | Compact Precision Blanace |
---|---|
Electrical Requirements | Universal Mains Adapter |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
17321683
|
Kern Sohn
PCB 1000-1 |
1000 g | |||||||
17311683
|
Kern Sohn
PCB 100-3 |
100 g |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17391683
|
Kern Sohn
PCB 3500-2 |
3500 g | |||||||
17381683
|
Kern Sohn
PCB 350-3 |
350 g |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
- Freely programmable weighing unit, e.g. display direct in special units such as length of thread g/m, paper weight g/m2, or similar
- With the recipe function you can weigh the different ingredients of a mixture. As a check, you can also call up the total weight of all the ingredients
- Plus/minus weighing procedures
- Protective working cover included with delivery
Specifications
Compact Precision Blanace | |
Universal Mains Adapter |