Learn More
Kern Sohn Compact Laboratory Balance PCB
The standard balance ideal for use in the laboratory.
Brand:  Kern Sohn PCB 350-3
Encompass_Preferred
 View more versions of this product
Product Code. 17381683
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
  • Freely programmable weighing unit, e.g. display direct in special units such as length of thread g/m, paper weight g/m2, or similar
  • With the recipe function you can weigh the different ingredients of a mixture. As a check, you can also call up the total weight of all the ingredients
  • Plus/minus weighing procedures
  • Protective working cover included with delivery
Specifications
Specifications
350 g
0.005 g
Compact Precision Blanace
81 mm
0.001 g
0.002 g
81 mm
Universal Mains Adapter