Learn More
Kern Sohn Compact Laboratory Balance PCB
The standard balance ideal for use in the laboratory.
Brand: Kern Sohn PCB 350-3
This item is not returnable.
View return policy
Description
- PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
- Freely programmable weighing unit, e.g. display direct in special units such as length of thread g/m, paper weight g/m2, or similar
- With the recipe function you can weigh the different ingredients of a mixture. As a check, you can also call up the total weight of all the ingredients
- Plus/minus weighing procedures
- Protective working cover included with delivery
Specifications
350 g | |
0.005 g | |
Compact Precision Blanace | |
81 mm |
0.001 g | |
0.002 g | |
81 mm | |
Universal Mains Adapter |