Learn More
Conductivity Solution, 20000μS.cm-1, Fisher Chemical™ Product Code.: 12954505
Conductivity Solution, 20000μS.cm-1, Fisher Chemical™
BIOPHARMA
Product Code. 12954505
500 mL
Click to view available options
Quantity:
500 mL
1000 mL
Product Code. 12954505
Brand: Fisher Chemical CTE 014 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Quantity 500 mL
Conductivity 20,000μS/cm
Type Conductivity Solution
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1037118 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberCTE 014 500
Lot Number1006070 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberCTE 014 500
Lot Number1010620 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberCTE 014 500
Lot Number1029606 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberCTE 014 500
Product Content Correction
Product Title
Conductivity Solution, 20000μS.cm-1, Fisher Chemical™ >