Learn More
MOLYKOTE™ High Vacuum Grease
For use with vacuum pumps.
Brand: Cole Parmer 79751-30
This item is not returnable.
View return policy
Description
For use with vacuum pumps
- Stiff, nonmelting silicone lubricant that maintains its consistency from –40 to 500°F
- Nonoxidizing and nongumming
- Has excellent resistance to water and chemicals
Specifications
Vacuum Grease | |
Light gray | |
-40°F to 932°F | |
5.29 oz. |
Vacuum Pumps | |
1 Ea. | |
-40°C to +500°C | |
150 g |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
MOLYKOTE™ High Vacuum Grease > Volume: 150g
Spot an opportunity for improvement?Share a Content Correction