Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Brand:  Corning™ 91-200-42
 View more versions of this product
Product Code. 17283574
  • 56.84 EUR / Each
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 L
EVA
1/Pk.
Each
Heat sealable flexible tubing
1/Ea.
Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site
Documents
One moment while we fetch your results.
Product Title
Corning™ Collection Bags > EVA; 1 L