Learn More
Corning™ Costar™ Centrifugal Devices: Spin-X™ LC
Handles volumes up to 500μL. Corning™ Costar™ Centrifugal Devices: Spin-X™ LC includes a nylon membrane for applications such as HPLC sample preparation.
Brand: Corning™ 8170
381.38 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
- Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Specifications
Nylon | |
Non-sterile | |
0.5 mL |
0.45 μm | |
Spin-X LC |
COR011 CORNING FILTRATION GUIDE
File size: 753 kB
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
54Hours
15Minutes
34Seconds
13New Offer Save Up to 92%
Product Content Correction
Product Title
Corning™ Costar™ Centrifugal Devices: Spin-X™ LC > Nylon; 0.45 μm; No
Spot an opportunity for improvement?Share a Content Correction