Learn More
Hirschmann™ Counting Chambers
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann Instruments™ Counting Chambers are available in a variety of depths to fit a range of systems.
67.18 EUR - 174.00 EUR
Specifications
Quantity 1Each
View More Specs
Products 4
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10315421
View Documents
Hirschmann™
8100204
Counting Chambers, Bright lined; Neubauer-improved
155.00 EUR
Each
10200872
View Documents
Hirschmann™
8100103
Counting Chambers, Neubauer
67.18 EUR
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
10527831
View Documents
Hirschmann™
8110204
Counting Chambers, Bright lined with clamps; Neubauer-improved
174.00 EUR
Each
10794873
View Documents
Hirschmann™
8100104
Counting Chambers, Neubauer-improved
68.33 EUR
Each
Estimated Shipment: 15-05-2025
Description
  • Double net ruling
  • Available bright-lined or no, with or without clamps
  • Includes 2 cover glasses for haemocytometer
  • CE acc. IVD 98/79 EC
Specifications
Specifications
1Each
Documents
Product Certifications
Product Content Correction
Product Title
Hirschmann™ Counting Chambers