Learn More
Shamrock™ Cryostick Labels
Brand:  Shamrock™ FCS-7515R
 View more versions of this product
Product Code. 11832661
  • 77.85 EUR / Pack of 1000
Specifications
Permanent
Cryostick
Cryogenic use
-80°C
19 x 38 mm
White
Cryogenic Label
Thermal printer
19 x 38 mm