Learn More
Sartorius Cubis™ II Analytical Lab Balance
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
The Cubis™ II Analytical Lab Balance has a maximum weighing capacity between 120 g and 520 g in combination with readability of 0.1 mg.
Brand: Sartorius MCA124S-2CEU-I QP99
15605.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The New Generation of Modular Premium Laboratory Balances
- Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
- One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
- All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
- These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
- It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Internal | |
D-sub connector, 9-pin | |
Color Touchscreen | |
Draft Shield I, QP99 | |
1 sec. | |
100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A |
Balance with Type Approval Certificate for China | |
120 g | |
0.1 mg | |
Aluminum, plastic PBT, Optiwhite float glass and stainless steel | |
Analytical Balance |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Sartorius Cubis analytical balances
Product Content Correction
Product Title
Sartorius Cubis™ II Analytical Lab Balance > Cubis™ II Analytical Lab Balance; Capacity: 120 g; Includes: Draft Shield I, QP99
Spot an opportunity for improvement?Share a Content Correction