Learn More
CUDC-101, MedChemExpress
MedChemExpress CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.
Brand: Medchem Express HY-10223
This item is not returnable.
View return policy
Description
CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.Specifications
CUDC-101 | |
White | |
C24H26N4O4 | |
DMSO : 25 mg/mL (57.54 mM; Need ultrasonic) | |
434.49 | |
97.0% | |
Solid |
1012054-59-9 | |
Research | |
5 mg | |
C#CC1=CC=CC(NC2=NC=NC3=CC(OC)=C(C=C23)OCCCCCCC(NO)=O)=C1 | |
434.49 | |
Research | |
Cancer-Kinase/protease |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
CUDC-101, MedChemExpress