missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isocyanide, 96%, Thermo Scientific Chemicals

180.90 EUR - 670.34 EUR

Chemical Identifiers

CAS 68498-54-4
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.14
InChI Key XTLSDEJNOQVCHL-UHFFFAOYSA-N
Synonym cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762Show MoreShow Less
PubChem CID 2734784
IUPAC Name isocyanocyclopentane
SMILES [C-]#[N+]C1CCCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10506983
SDS View Documents
Thermo Scientific Acros
372090010
1 g Glass bottle
180.90 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
11985701
View Documents
Thermo Scientific Acros
372090050
5 g Glass bottle
670.34 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentyl isocyanide, 96%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.