missing translation for 'onlineSavingsMsg'
Learn More
Cyclopropanesulfonamide, 97%, Thermo Scientific™ Product Code.: 15441788

Cyclopropanesulfonamide, 97%, Thermo Scientific™

Product Code. 15441788
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15441788

Brand: Thermo Scientific Alfa Aesar H55507.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cyclopropanesulfonamide is used as fine chemicals to the pharmaceutical, agrochemical, aroma, polymer and electronic industries.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154350-28-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
MDL Number MFCD08705286
InChI Key WMSPXQIQBQAWLL-UHFFFAOYSA-N
Synonym cyclopropanesulfonyl amide, cyclopropanesulphonamide, cyclopropyl sulfonamide, cyclopropane sulfonamide, cyclopropanesulfonic acid amide, cyclopropylsufonamide, cyclopropysulfonamide, sulphamoylcyclopropane, cyclopropylsulphonamide, cyclopropyl-sulfonamideShow MoreShow Less
PubChem CID 15765418
IUPAC Name cyclopropanesulfonamide
SMILES C1CC1S(=O)(=O)N

Specifications

Melting Point 101°C to 106°C
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 121.15
Percent Purity 97%
Chemical Name or Material Cyclopropanesulfonamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBF160501 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberX04H056 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopropanesulfonamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.