Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket, Threaded
Cylindrical shape, with graduations
1250.00 EUR - 1630.00 EUR
Specifications
Description | DURAN™ Tubing |
---|---|
Includes | Laboratory Flange 150 |
Product Type | Reaction vessel |
Shape | Cylindrical |
Bottom Shape | Flat |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10648170
|
Lenz Laborglasinstrumente™
06010473 |
2 L |
1250.00 EUR
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11522244
|
Lenz Laborglasinstrumente™
06010476 |
3 L |
1300.00 EUR
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11542244
|
Lenz Laborglasinstrumente™
06010482 |
5 L |
1455.00 EUR
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11552244
|
Lenz Laborglasinstrumente™
06010485 |
6 L |
1630.00 EUR
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
Description
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.Specifications
DURAN™ Tubing | |
Reaction vessel | |
Flat |
Laboratory Flange 150 | |
Cylindrical | |
190 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket, Threaded