missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYSLTR2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577473
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Receptor for cysteinyl leukotrienes. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. Stimulation by BAY u9773, a partial agonist, induces specific contractions of pulmonary veins and might also have an indirect role in the relaxation of the pulmonary vascular endothelium. The rank order of affinities for the leukotrienes is LTC4 = LTD4 >> LTE4.Specifications
CYSLTR2 (extracellular) | |
Polyclonal | |
Unconjugated | |
CYSLTR2 | |
CysLTR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57105 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunofluorescence, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q9NS75 | |
CYSLTR2 | |
Peptide (C)HLTTWKVGL(S)KDRLH, corresponding to amino acid residues 270-284 of human CysLTR2 | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CYSLTR2 (extracellular) Polyclonal Antibody, Invitrogen™ > 50 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction