missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (97A1015) - (cleavage specific) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 16 publications
207.00 EUR - 452.00 EUR
Specifications
Antigen | D4-GDI/RhoGDI2 |
---|---|
Clone | 97A1015 |
Concentration | 1.0 mg/ml |
Dilution | Western Blot 2 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) 0.5-1 ug/10^6 cells |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18026094
|
Bio-Techne
NB100-56725SS |
0.025 mg |
207.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747583
|
Novus Biologicals
NB100-56725 |
0.1 mg |
452.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
D4-GDI/RhoGDI2 | |
1.0 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse | |
ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
ARHGDIB | |
IgG2b κ | |
Protein G purified | |
In anti-Fas treated Jurkat, an approx. 23 kDa band should be observed. |
97A1015 | |
Western Blot 2 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) 0.5-1 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
P52566 | |
397 | |
A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human D4-GDI/RhoGDI2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D4-GDI/RhoGDI2 Antibody (97A1015) - (cleavage specific) - BSA Free, Novus Biologicals™