missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl trisulfide, 98.8%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g
Specifications
Specifications
Color | Colorless |
Assay Percent Range | 0.988 |
Quantity | 25 mg |
Formula Weight | 178.3 |
Physical Form | Liquid |
Chemical Name or Material | DATS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diallyl trisulfide, 98.8%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction