Learn More
Bel-Art™ SP Scienceware™ Dialysis Bag Clip Holders
Strong support for suspending dialysis bags in beakers or pitchers
Brand: Bel-Art™ SP Scienceware™ F18237-0000
27.08 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suspend dialysis bags in large beakers or pitchers up to 0.63cm (0.25 in.) thick with these strong clips
- Bags slide easily into slot and then holder can be clipped onto the container’s edge
- Smooth polypropylene surface may be marked with identifying sequence numbers
- Autoclavable
Specifications
Dialysis Bag Clips | |
Autoclavable | |
Natural | |
Non-sterile |
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Dialysis Bag Clip Holders > Dialysis Bag Clip Holder
Spot an opportunity for improvement?Share a Content Correction