missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: MacConkey Agar
For differential isolation of enteric bacilli based on lactose fermentation
403.00 EUR
Specifications
Content And Storage | Refrigerate |
---|---|
Form | Dehydrated |
Media Type | MacConkey Agar |
Product Type | Agar |
Formulation | Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
Product Code | Brand | Quantity | Packaging Type | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Certifications/Compliance | Price | Quantity & Availability | |||
10537884
|
BD
212122 |
2 kg | Pail | CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP |
403.00 EUR
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Specifications
Refrigerate | |
MacConkey Agar | |
Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
Dehydrated | |
Agar | |
For differential isolation of enteric Bacilli based on Lactose fermentation |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MacConkey Agar
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H371-May cause damage to organs.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : bile salts
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: MacConkey Agar