Learn More
SCHOTT AG Lighting and Imaging™ Diffusor for LED Ring Light
The Diffusor for LED Ring Light avoids LED reflections. For use with VisLED Brightfield Ringlights, EasyLED Ringlight and EasyLED Ringlight Plus.
Brand:  SCHOTT AG Lighting and Imaging™ 400570
Product Code. 12836553
  • 310.00 EUR / Each
Estimated Shipment: 23-04-2025
Description
  • Prevents LED reflections.
  • Includes adapting thread M110 x 1.
  • Fits 66 mm ID.
Specifications
Specifications
Diffusor
66 mm
EasyLED Ringlights or VisiLED Ringlights S80-25 or S80-55
2.6 in.
Stereo Microscopy Illumination
Documents
One moment while we fetch your results.
Product Title
SCHOTT AG Lighting and Imaging™ Diffusor for LED Ring Light > Diffusor; For Use With: 66 mm ID LED Ringlight