missing translation for 'onlineSavingsMsg'
Learn More
Dihydrocoumarin, 99%, Thermo Scientific Chemicals Product Code.: 11414747

Dihydrocoumarin, 99%, Thermo Scientific Chemicals

Product Code. 11414747
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11414747

Brand: Thermo Scientific Alfa Aesar B21229.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-84-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006881
InChI Key VMUXSMXIQBNMGZ-UHFFFAOYSA-N
Synonym dihydrocoumarin, 3,4-dihydrocoumarin, hydrocoumarin, chroman-2-one, benzodihydropyrone, melilotin, melilotol, 2-chromanone, 1,2-benzodihydropyrone, melilotic lactone
PubChem CID 660
ChEBI CHEBI:16151
IUPAC Name 3,4-dihydrochromen-2-one
SMILES C1CC(=O)OC2=CC=CC=C21

Specifications

Melting Point 24°C to 25°C
Density 1.169
Boiling Point 272°C
Flash Point 130°C (266°F)
Refractive Index 1.556
Quantity 500 g
Beilstein 4584
Formula Weight 148.16
Percent Purity 99%
Chemical Name or Material Dihydrocoumarin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10191534 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberB21229.0E,B21229.36,B21229.22
Lot Number10244580 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog NumberB21229.0E,B21229.36,B21229.22
Lot Number10166246 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21229.0E,B21229.36,B21229.22
Lot Number10146619 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21229.0E,B21229.36,B21229.22
Lot Number10191534 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21229.0E,B21229.36,B21229.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dihydrocoumarin, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.