missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyanodithioiminocarbonate, 95%

Product Code. 11408883
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11408883

Brand: Thermo Scientific Alfa Aesar A11043.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10191-60-3 | C4H6N2S2 | 146.226 g/mol

Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

Solubility
Slightly soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-60-3
Molecular Formula C4H6N2S2
Molecular Weight (g/mol) 146.226
MDL Number MFCD00009825
InChI Key IULFXBLVJIPESI-UHFFFAOYSA-N
Synonym dimethyl cyanocarbonimidodithioate, dimethyl cyanodithioiminocarbonate, dimethyl n-cyanodithioiminocarbonate, carbonimidodithioic acid, cyano-, dimethyl ester, s,s-dimethyl cyanoimidodithiocarbonate, cyanoimino bis methylthio methane, dimethyl n-cyanoimidodithiocarbonate, dimethyl cyanimidodithiocarbonate, dimethyl cyanoiminodithiocarbonate, dimethyl n-cyanodithiocarbonimidate
PubChem CID 66289
IUPAC Name bis(methylsulfanyl)methylidenecyanamide
SMILES CSC(=NC#N)SC

Specifications

Melting Point 50°C to 56°C
Flash Point >110°C (230°F)
Odor Stench
Linear Formula NCN=C(SCH3)2
Quantity 100 g
UN Number UN2923
Beilstein 635998
Solubility Information Slightly soluble in methanol.
Formula Weight 146.23
Percent Purity 95%
Chemical Name or Material Dimethyl cyanodithioiminocarbonate
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis