missing translation for 'onlineSavingsMsg'
Learn More
Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%, Thermo Scientific Chemicals Product Code.: 10138752

Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%, Thermo Scientific Chemicals

Product Code. 10138752
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10138752

Brand: Thermo Scientific Acros 382400050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118612-00-3
Molecular Formula C32H12BF20N
Molecular Weight (g/mol) 801.23
MDL Number MFCD01074420
InChI Key BRHZQNMGSKUUMN-UHFFFAOYSA-O
Synonym dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
PubChem CID 10996402
IUPAC Name dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1

Specifications

Color Blue-White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 801.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dimethylanilinium tetrakis (pentafluorophenyl)borate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethylanilinium tetrakis (pentafluorophenyl)borate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.