Learn More
Sartorius™ Disposable Sample Pans
Use with Sartorius™ Moisture Balances. Sartorius™ Disposable Aluminum Sample Pans provide easy clean-up when determining moisture content.
Brand: Sartorius™ 6965542
45.89 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Use with Sartorius™ Moisture Balances. Sartorius™ Disposable Aluminum Sample Pans provide easy clean-up when determining moisture content.
- Aluminum
Specifications
Flat | |
Aluminum | |
Balance Pan |
Single-use | |
Circular | |
90 mm |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%