Learn More
Epredia™ Disposable Blade Carriers
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Brand: Epredia™ 705050
This item is not returnable.
View return policy
Description
Disposable Blade Carriers- Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
- Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
- Quick clamp system for high- and low-profile blades
- Adjustable clearance angle
- Moveable knife guard covers the blade entirely in any position
- Knife carrier base included
Specifications
E Type blade carrier for dovetail style units | |
Disposable Blade Carrier E | |
Dovetail Style Unit |
Product Content Correction
Product Title
Epredia™ Disposable Blade Carriers > Disposable blade carrier
Spot an opportunity for improvement?Share a Content Correction