Learn More
Nexcelom™ Disposable Hemacytometer
Plastic cell counting chamber slides with 2 chambers per slide containing a hemacytometer grid on the slide
Brand: Nexcelom™ CP2-001
This item is not returnable.
View return policy
Description
- An enclosed chamber with two ports for sample introduction
- The chamber contains precisely spaced lines in a grid pattern
- There are two counting chambers per slide for replicates
- The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
Specifications
20 microliters | |
25 slides for 50 counts |