Learn More
Fisherbrand™ Disposable Inoculating Loops
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
163.00 EUR
Specifications
Material | Polystyrene |
---|---|
Disposable | Single-use |
Autoclavable | Not Autoclavable |
Product Code | Brand | Sterility | Volume | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Volume | Quantity | Price | Quantity & Availability | ||||
17783667
|
Fisherbrand™
01189165 |
Sterile | 1 μL | Cs of 250 (5 Pks. of 50/ Individually Packed) |
163.00 EUR
Pack of 250 |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||
17793667
|
Fisherbrand™
01189169 |
Sterile | 10 μL | Cs of 125 (5 Pks. of 25/ Individually Packed) |
163.00 EUR
Pack of 125 |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||
Description
- Ideal for anaerobic microbiology applications where deep agar inoculation is required
- Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
- High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
- Rigid enough to penetrate the media for subsurface inoculation
- Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
- Can be used under hoods without danger
- Color-coded for ease of size identification
Specifications
Polystyrene | |
Single-use | |
Not Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Disposable Inoculating Loops