missing translation for 'onlineSavingsMsg'
Learn More
DL-SulForaphane, 97%, MP Biomedicals™ Product Code.: 11492371

DL-SulForaphane, 97%, MP Biomedicals™

Product Code. 11492371 Shop All MP Biomedicals Products
25 mg
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy

Product Code. 11492371

Brand: MP Biomedicals 0219378125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 4478-93-7
Color Yellow
Assay Percent Range 0.97
Molecular Formula C6H11NOS2
MDL Number MFCD00198068,MFCD09039283
Quantity 25 mg
Synonym sulforaphane, dl-sulforaphane, sulforafan, 1-isothiocyanato-4-methylsulfinyl butane, d,l-sulforaphane, sulforaphane unspecified, r,s-sulforaphane, ccris 7221, 1-isothiocyanato-4-methylsulfinyl-butane, butane, 1-isothiocyanato-4-methylsulfinyl
InChI Key SUVMJBTUFCVSAD-UHFFFAOYNA-N
SMILES CS(=O)CCCCN=C=S
IUPAC Name 1-isothiocyanato-4-methanesulfinylbutane
Molecular Weight (g/mol) 177.28
PubChem CID 5350
ChEBI CHEBI:47807
Formula Weight 177.3
Physical Form Liquid
Chemical Name or Material DL-Sulforaphane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-SulForaphane, 97%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.