Learn More
Thermo Scientific™ DNA Marker, broad range
75.98 EUR - 390.95 EUR
Specifications
Product Type Molecular Weight Marker
Form Liquid
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15454699
View Documents
Thermo Scientific™
J62291.XF
1 mL
75.98 EUR
1mL
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
15444699
View Documents
Thermo Scientific™
J62291.S0
2 mL
390.95 EUR
2mL
Estimated Shipment: 10-02-2025
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Molecular Weight Marker