missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553208
This item is not returnable.
View return policy
Description
Immunogen sequence: FHDCEDAARE LQSQIQEARA DLARR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 92%.
Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. The protein encoded by this gene is a subunit of dolichyl-phosphate mannosyltransferase and acts as a stabilizer subunit of the dolichyl-phosphate mannosyltransferase complex.
Specifications
DPM3 | |
Polyclonal | |
Unconjugated | |
DPM3 | |
CDG1O; dolichol-phosphate mannose synthase subunit 3; Dolichol-phosphate mannosyltransferase subunit 3; dolichyl-phosphate beta-D-mannosyltransferase subunit 3; dolichyl-phosphate mannosyltransferase polypeptide 3; dolichyl-phosphate mannosyltransferase subunit 3; DPM synthase complex subunit 3; DPM synthase subunit 3; DPM3; Mannose-P-dolichol synthase subunit 3; MGC125904; MGC125905; MGC34275; MPD synthase subunit 3; prostin 1; prostin-1; testicular tissue protein Li 58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54344 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2X0 | |
DPM3 | |
Recombinant protein corresponding to Human DPM3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DPM3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Spot an opportunity for improvement?Share a Content Correction