Description
- Shade 5 lens is ideal for welding applications
- Low-profile design and toric lens for a large panoramic field of vision
- OTG: fits comfortably over most prescription spectacles
- Can be worn comfortably with half-mask respirator
- Soft, elastomer body flexes and conforms to the face for a comfortable, gap-free fit
- Indirect venting system and Dura-streme coating to minimize fogging
- Anti-fog and scratch resistant
- Large, adjustable headband
- Easy, economical lens replacement
Specifications
Specifications
Lens Type | Dura-streme |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | IR/UV 5.0 |
For Use With (Application) | Welding, industry, construction, maintenance |
Frame Color | Blue, Gray |
Headband Material | Neoprene |
Lens Material | Polycarbonate |
Frame Material | PP/TPE |
Product Content Correction
Product Title
Honeywell™ Duramaxx Safety Goggles, IR Shade 5.0 >
Spot an opportunity for improvement?Share a Content Correction