Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
2-way tap plug with thread.
1055.00 EUR - 1160.00 EUR
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
15864518
|
DWK Life Sciences
286036706 |
14.5 NS |
1055.00 EUR
Pack of 50 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
15884518
|
DWK Life Sciences
286037308 |
18.8 NS |
1160.00 EUR
Pack of 50 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Description
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
- With thread, ground
- 2-way tap plug with 2 almost parallel inclined holes
- massive tap plug
Specifications
Borosilicate Glass (3.3) | |
Autoclavable | |
Installation in glass apparatus and devices |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground