Learn More
Abeba™ Dynamic 37361 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37361/38
 View more versions of this product
Product Code. 15671018
  • 76.65 EUR / 1 pair
Estimated Shipment: 18-02-2025
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
37361
Black
38
Polyurethane