Learn More
Abeba™ Dynamic 37361 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
76.65 EUR
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15641018
|
Abeba™
37361/35 |
35 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15651018
|
Abeba™
37361/36 |
36 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15661018
|
Abeba™
37361/37 |
37 |
76.65 EUR
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15671018
|
Abeba™
37361/38 |
38 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15681018
|
Abeba™
37361/39 |
39 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15691018
|
Abeba™
37361/40 |
40 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15601028
|
Abeba™
37361/41 |
41 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15611028
|
Abeba™
37361/42 |
42 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15621028
|
Abeba™
37361/43 |
43 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15631028
|
Abeba™
37361/44 |
44 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15641028
|
Abeba™
37361/45 |
45 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15651028
|
Abeba™
37361/46 |
46 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15661028
|
Abeba™
37361/47 |
47 |
76.65 EUR
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15671028
|
Abeba™
37361/48 |
48 |
76.65 EUR
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Polyurethane |
Black | |
Slip-resistant | |
37361 |