Learn More
(E)-[6]-Dehydroparadol, MedChemExpress
MedChemExpress (E)-[6]-Dehydroparadol, an oxidative metabolite of [6]-Shogaol (HY-14616), is a potent Nrf2 activator. (E)-[6]-Dehydroparadol can inhibit the growth and induce the apoptosis of human cancer cells.
Brand: Medchem Express HY-77293
This item is not returnable.
View return policy
Description
(E)-[6]-Dehydroparadol, an oxidative metabolite of [6]-Shogaol (HY-14616), is a potent Nrf2 activator. (E)-[6]-Dehydroparadol can inhibit the growth and induce the apoptosis of human cancer cells.Specifications
(E)-[6]-Dehydroparadol | |
Orange | |
C17H24O3 | |
DMSO : ≥ 140 mg/mL (506.57 mM) | |
276.37 | |
95.0% | |
Solid |
878006-06-5 | |
Research | |
100 mg | |
CCCCCCCC(/C=C/C1=CC=C(O)C(OC)=C1)=O | |
276.37 | |
Research |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
(E)-[6]-Dehydroparadol, MedChemExpress