Description
- These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
- Test plugs can not be used for hearing protection
- Probed test earplug for use with 3M™ E-A-Rfit™ Validation System
Specifications
Specifications
Color | Yellow |
Size | Small |
Packaging Type | Polybag |
Pairs per Package | 50 Pairs/Box |
Corded | Uncorded |
Plug Shape | Cylinder |
Dielectric | No |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Classic™ Probed Test Plugs >
Spot an opportunity for improvement?Share a Content Correction