Learn More
Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 0.8% (EU only)
E-Gel™ Agarose Gels Starter Pak, 0.8% (EU only)
Brand: Invitrogen™ G600008EU
This item is not returnable.
View return policy
Description
E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
Specifications
E-Gel™ Agarose Gels Starter Pak, 0.8%, E-Gel™ Gel Type, Ethidium Bromide In-Gel Dye, 12 Wells, Single Comb Well Format, Store at Room Temperature, For Nucleic Acid Gel Electrophoresis and Blotting | |
Agarose Gels Starter Kit | |
6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual, 18-packs contain 18 gels and a manual | |
Room temperature |
0.8% | |
DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis | |
Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 0.8% (EU only) > 1 pack
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction