Learn More
STEMCELL Technologies EasySep™ HLA B Cell Enrichment: Complete Processing Kit for Whole Blood
Immunomagnetic isolation of untouched human B cells
Brand: STEMCELL Technologies 19954HLA
This item is not returnable.
View return policy
Description
- The EasySep™ HLA B Cell Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human B cells from HetaSep™-treated whole blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD33, CD36, CD41, CD43, CD56, CD66b, glycophorin A and dextran-coated magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
B Cells | |
Cell Isolation | |
200 mL whole blood |