Learn More
Fisherbrand™ Ebony Treatment Dish
Protects DNA from damage. Fisherbrand™ Ebony Treatment Dishes are manufactured with a darkened acrylic to reduce exposure to light that may be harmful to DNA.
Brand: Fisherbrand™ STAINDISH
Description
- Manufactured from Ebony Acrylic
- Minimizes exposure to light and reduces background DNA damage
Specifications
Ebony | |
Comet Assay Tank | |
Acrylic | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Fisherbrand Comet assay tanks
File size: 1.4 MB
Product Content Correction
Product Title
Fisherbrand™ Ebony Treatment Dish >
Spot an opportunity for improvement?Share a Content Correction