Learn More
Labcon™ Eclipse™ 200 μL Beveled Point Yellow Pipette Tips
These pipette tips fit most popular 200 μL pipettors.
Brand: Labcon™ 1030-790-006-9
54.39 EUR valid until 2025-12-31
Use promo code "24311" to get your promotional price.
Description
- Beveled point
- ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Polypropylene USP Class VI Resin
- Made in USA
Specifications
200 μL | |
Non-sterile | |
Purple | |
1.95 in. | |
ISO 9001:2015, FDA CFR21 GMP, USP Class VI | |
Liquid Handling | |
Beveled | |
Single-use | |
No | |
Polypropylene Resin |
Non-filtered | |
Yellow | |
Autoclavable | |
49.7 mm | |
ATP, RNase/Dnase, Endotoxin-free | |
Stack Rack | |
192 Tips/Rack, 5 Racks/Pk., 10 Pk./Cs. | |
Universal Fit | |
Eclipse |